• USA Home
  • Search Results

  • isoamylpropionate1442110568011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoamyl propionate

3 Products
Synonym:   Amyl propionate
Image of Isoamyl propionate
CAS Number:
Linear Formula:
 C2H5CO2CH2CH2CH(CH3)2
Molecular Weight:
 144.21
 Council of Europe no.:
 417
 EC Number:
 FEMA Number:
 2082
 Flavis number:
 9.136
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Potency
  Assay
  Special Grade
  Agency/Method
Show More