• USA Home
  • Search Results

  • isobutylcinnamate2042612267811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isobutyl cinnamate

2 Products
Synonym:   2-Methylpropyl cinnamate, 2-methylpropyl 3-phenylpropenoate
Image of Isobutyl cinnamate
CAS Number:
Linear Formula:
 C6H5CH=CHCO2CH2CH(CH3)2
Molecular Weight:
 204.26
 Council of Europe no.:
 327
 EC Number:
 FEMA Number:
 2193
 Flavis number:
 9.734
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More