• USA Home
  • Search Results

  • isobutylmethylketone1001610810111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isobutyl methyl ketone

2 Products
Synonym:   4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone
Image of Isobutyl methyl ketone
CAS Number:
Linear Formula:
 (CH3)2CHCH2COCH3
Molecular Weight:
 100.16
 Beilstein/REAXYS Number:
 605399
 EC Index Number:
 203-550-1
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
  Physical Form
  pH Value
  Transition temperature
  Assay
Show More