• USA Home
  • Search Results

  • isobutyricanhydride158199772311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isobutyric anhydride

3 Products
Synonym:   2-Methylpropionic anhydride
Image of Isobutyric anhydride
CAS Number:
Linear Formula:
 (CH3)2CHCO2COCH(CH3)2
Molecular Weight:
 158.19
 Beilstein/REAXYS Number:
 386267
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More