• USA Home
  • Search Results

  • isofraxidin2221948621511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isofraxidin

2 Products
Synonym:   6,8-Dimethoxyumbelliferone, 7-Hydroxy-6,8-dimethoxychromen-2-one, 7-Hydroxy-6,8-dimethoxycoumarin
Image of Isofraxidin
CAS Number:
Empirical Formula (Hill Notation):
 C11H10O5
Molecular Weight:
 222.19
 Beilstein/REAXYS Number:
 202652
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More