• USA Home
  • Search Results

  • isoimperatorin2702848245111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoimperatorin

2 Products
Synonym:   4-(3-Methylbut-2-enyloxy)furo[3,2-g]chromen-7-one, 4-[(3-Methyl-2-buten-1-yl)oxy]-7H-furo[3,2-g][1]benzopyran-7-one
Image of Isoimperatorin
CAS Number:
Empirical Formula (Hill Notation):
 C16H14O4
Molecular Weight:
 270.28
 Beilstein/REAXYS Number:
 1291723
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More