• USA Home
  • Search Results

  • isopentanesolution72157878411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopentane Solution

1 Products
Synonym:   2-Methylbutane, Isopentane
Image of Isopentane Solution
CAS Number:
Linear Formula:
 CH3CH2CH(CH3)2
Molecular Weight:
 72.15
 Beilstein/REAXYS Number:
 1730723
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More