• USA Home
  • Search Results

  • isophthalaldehyde1341362619711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isophthalaldehyde

1 Products
Synonym:   Benzene-1,3-dicarboxaldehyde
Image of Isophthalaldehyde
CAS Number:
Linear Formula:
 C6H4-1,3-(CHO)2
Molecular Weight:
 134.13
 Beilstein/REAXYS Number:
 1561038
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More

Product #
Description
115282 Isophthalaldehyde, 97%,