• USA Home
  • Search Results

  • isophthalicacid1661312191511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isophthalic acid

2 Products
Synonym:   Benzene-1,3-dicarboxylic acid
Image of Isophthalic acid
CAS Number:
Linear Formula:
 C6H4-1,3-(CO2H)2
Molecular Weight:
 166.13
 Beilstein/REAXYS Number:
 1909332
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More