• USA Home
  • Search Results

  • isophthaloylchloride203029963811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isophthaloyl chloride

2 Products
Synonym:   Isophthaloyl dichloride
Image of Isophthaloyl chloride
CAS Number:
Linear Formula:
 C6H4-1,3-(COCl)2
Molecular Weight:
 203.02
 Beilstein Registry Number:
 638342
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More