• USA Home
  • Search Results

  • isopropanol123456763011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopropanol

2 Products
Synonym:   2-Propanol solution, 2-Propanol, IPA, Isopropanol, Isopropyl alcohol
Image of Isopropanol
CAS Number:
 MDL number:
 NACRES:
 NA.05
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More