• USA Home
  • Search Results

  • isopropylalcohol60106763011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopropyl alcohol

8 Products
Synonym:   2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol
Image of Isopropyl alcohol
CAS Number:
Linear Formula:
 (CH3)2CHOH
Molecular Weight:
 60.10
 Beilstein/REAXYS Number:
 635639
 Council of Europe no.:
 4146
 EC Number:
 FEMA Number:
 2929
 Flavis number:
 2.079
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Absorption
  Document Links
  Allergen
  Organoleptic property
  Color
  Biological Source
  Vapor pressure
  Transition temperature
  Application(s)
  Ice Shipping
  Physical Form
  pH Value
  Agency/Method
  Special Grade
Show More