• USA Home
  • Search Results

  • isopropylcinnamate19024778006511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopropyl cinnamate

1 Products
Synonym:   Isopropyl 3-phenylpropenoate
Image of Isopropyl cinnamate
CAS Number:
Linear Formula:
 C6H5CH=CHCO2CH(CH3)2
Molecular Weight:
 190.24
 Council of Europe no.:
 325
 EC Number:
 FEMA Number:
 2939
 Flavis number:
 9.732
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More