• USA Home
  • Search Results

  • isopropylisothiocyanate10117225373811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopropyl isothiocyanate

1 Products
Synonym:   Isopropyl mustard oil
Image of Isopropyl isothiocyanate
CAS Number:
Linear Formula:
 (CH3)2CHNCS
Molecular Weight:
 101.17
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More