• USA Home
  • Search Results

  • isopropylpalmitate2985014291611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isopropyl palmitate

4 Products
Synonym:   Isopropyl hexadecanoate
Image of Isopropyl palmitate
CAS Number:
Linear Formula:
 CH3(CH2)14COOCH(CH3)2
Molecular Weight:
 298.50
 Beilstein/REAXYS Number:
 1786567
 EC Number:
 Flavis number:
 9.606
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Assay
Show More