• USA Home
  • Search Results

  • isorhamnetin3rutinoside6245460480811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isorhamnetin 3-rutinoside

1 Products
Synonym:   3,4′,5,7-Tetrahydroxy 3′-methoxyflavone 3-rutinoside, 3-Rutinosylisorhamnetin, 3-O-Methylquercetin 3-rutinoside, Isorhamnetin 3-O-rutinoside, Isorhamnetin 3-rhamnoglucoside, Isorhamnetol 3-rutinoside, Narcissin, Narcissoside, Quercetin 3′-methyl ether 3-rutinoside
Image of Isorhamnetin 3-rutinoside
CAS Number:
Empirical Formula (Hill Notation):
 C28H32O16
Molecular Weight:
 624.54
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More