• USA Home
  • Search Results

  • isosilybinb4824414279622311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isosilybin B

2 Products
Synonym:   (2R,3R)-3,5,7-Trihydroxy-2-[(2S,3S)-2-(4-hydroxy-3-methoxyphenyl)-3-(hydroxymethyl)-2,3-dihydrobenzo[1,4]dioxin-6-yl]-4-chromanone
Image of Isosilybin B
CAS Number:
Empirical Formula (Hill Notation):
 C25H22O10
Molecular Weight:
 482.44
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More