• USA Home
  • Search Results

  • isovaleraldehyde861359086311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isovaleraldehyde

4 Products
Synonym:   3-Methylbutyraldehyde, 3-Methylbutanal, NSC 404119
Image of Isovaleraldehyde
CAS Number:
Linear Formula:
 (CH3)2CHCH2CHO
Molecular Weight:
 86.13
 Beilstein/REAXYS Number:
 773692
 Council of Europe no.:
 94
 EC Number:
 FEMA Number:
 2692
 Flavis number:
 5.006
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Document Links
  Allergen
  Application(s)
  Solubility
  Special Grade
  Organoleptic property
  Assay
Show More