• USA Home
  • Search Results

  • isovalericacid1021350374211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isovaleric acid

4 Products
Synonym:   3-Methylbutanoic acid, 3-Methylbutyric acid
Image of Isovaleric acid
CAS Number:
Linear Formula:
 (CH3)2CHCH2COOH
Molecular Weight:
 102.13
 Beilstein/REAXYS Number:
 1098522
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More