• USA Home
  • Search Results

  • isoxaflutole3593214111229011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoxaflutole

1 Products
Synonym:   5-Cyclopropyl-4-[2-(methylsulfonyl)-4-(trifluoromethyl)benzoyl]isoxazole
Image of Isoxaflutole
CAS Number:
Empirical Formula (Hill Notation):
 C15H12F3NO4S
Molecular Weight:
 359.32
 Beilstein Registry Number:
 8344543
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More