• USA Home
  • Search Results

  • isoxanthohumol3544052148211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoxanthohumol

4 Products
Synonym:   2,3-Dihydro-7-hydroxy-2-(4-hydroxyphenyl)-5-methoxy-8-(3-methyl-2-butenyl)-4H-1-benzopyran-4-one, 4,7-Dihydroxy-5-methoxy-8-prenylflavanone, Humulol, IXN, Isoxanthohumol
Image of Isoxanthohumol
CAS Number:
Empirical Formula (Hill Notation):
 C21H22O5
Molecular Weight:
 354.40
 Beilstein/REAXYS Number:
 10199264
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Physical Form
  Special Grade
  Application(s)
  Manufacturer designation
Show More