• USA Home
  • Search Results

  • isradipine371397569593111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isradipine

4 Products
Synonym:   4-(4-Benzofurazanyl)-1,-4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid methyl 1-methylethyl ester
Image of Isradipine
CAS Number:
Empirical Formula (Hill Notation):
 C19H21N3O5
Molecular Weight:
 371.39
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Physical Form
  Special Grade
Show More