• USA Home
  • Search Results

  • jasminabsolute12345802296611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Jasmin absolute

1 Products
Synonym:   Jasmine oil
CAS Number:
 FEMA Number:
 2598
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
Show More

Product #
Description
W259802 Jasmin absolute, FG,