• USA Home
  • Search Results

  • jodfenphos413001818170911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Jodfenphos

1 Products
Image of Jodfenphos
CAS Number:
Empirical Formula (Hill Notation):
 C8H8Cl2IO3PS
Molecular Weight:
 413.00
 Beilstein/REAXYS Number:
 1885795
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More