• USA Home
  • Search Results

  • juniperberryoil12345800268411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Juniper berry oil

1 Products
CAS Number:
 FEMA Number:
 2604
 MDL number:
 NACRES
 NA.21
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Geographic origin
  Biological Source
  Agency/Method
Show More

Product #
Description
W260401 Juniper berry oil, FG,