• USA Home
  • Search Results

  • kaempferitrin5785248238211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kaempferitrin

2 Products
Synonym:   3,7-Bis((6-deoxy-α-L-mannopyranosyl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Kaempferol 3,7-dirhamnoside, Lespedin, Lespenefril, Lespenephryl
Image of Kaempferitrin
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O14
Molecular Weight:
 578.52
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More