• USA Home
  • Search Results

  • kaempferol2862452018311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kaempferol

6 Products
Synonym:   3,4′,5,7-Tetrahydroxyflavone, 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Robigenin
Image of Kaempferol
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O6
Molecular Weight:
 286.24
 Beilstein/REAXYS Number:
 304401
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Shelf Life
  Physical Form
  Special Grade
  Titertest
  Solubility
Show More