• USA Home
  • Search Results

  • kaempferol3glucoside4483848010411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kaempferol 3-glucoside

3 Products
Synonym:   3,4′,5,7-Tetrahydroxyflavone 3-glucoside, 3-(β-D-Glucopyranosyloxy)-5,7-dihydroxy-2-(4-methoxyphenyl)-4H-1-benzopyran-4-one, 3-Glucosylkaempferol, Astragalin, Kaempferol 3-β-D-glucopyranoside, Kaempferol 3-glucoside
Image of Kaempferol 3-glucoside
CAS Number:
Empirical Formula (Hill Notation):
 C21H20O11
Molecular Weight:
 448.38
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Application(s)
  Manufacturer designation
  Assay
Show More