• USA Home
  • Search Results

  • kaempferol3rutinoside594521765084911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kaempferol 3-rutinoside

1 Products
Synonym:   3,4′,5,7-Tetrahydroxyflavone 3-rhamnoglucoside, Kaempferol 3-O-β -rutinoside, Nicotiflorin
Image of Kaempferol 3-rutinoside
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O15
Molecular Weight:
 594.52
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More