• USA Home
  • Search Results

  • kaempferol3sambubioside580492766151411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kaempferol 3-sambubioside

1 Products
Synonym:   Kaempferol 3-sambubioside, Leucoside
Image of Kaempferol 3-sambubioside
CAS Number:
Empirical Formula (Hill Notation):
 C26H28O15
Molecular Weight:
 580.49
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More