• USA Home
  • Search Results

  • kanamycinsulfatestreptomyceskanamyceticus582582538994011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kanamycin sulfate Streptomyces kanamyceticus

1 Products
Synonym:   Kanamycin A, Kanamycin sulfate from Streptomyces kanamyceticus, Kanamycin sulfate salt
Image of Kanamycin sulfate Streptomyces kanamyceticus
CAS Number:
Empirical Formula (Hill Notation):
 C18H36N4O11 · H2O4S
Molecular Weight:
 582.58
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More