• USA Home
  • Search Results

  • ketaminehydrochloride27419186766911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-Ketamine hydrochloride

3 Products
Synonym:   (±)-2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride
Image of (±)-Ketamine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C13H16ClNO · HCl
Molecular Weight:
 274.19
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More