• USA Home
  • Search Results

  • kojicacid1421150130411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kojic acid

2 Products
Synonym:   2-Hydroxymethyl-5-hydroxy-γ-pyrone, 5-Hydroxy-2-hydroxymethyl-4H-4-pyranone
Image of Kojic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H6O4
Molecular Weight:
 142.11
 Beilstein/REAXYS Number:
 120895
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More