• USA Home
  • Search Results

  • kovacsreagentforindoles1491910010711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kovac’s reagent for indoles

2 Products
Synonym:   4-(Dimethylamino)benzaldehyde solution, Indole reagent according to Kovac
Image of Kovac’s reagent for indoles
CAS Number:
Empirical Formula (Hill Notation):
 C9H11NO
Molecular Weight:
 149.19
 Beilstein/REAXYS Number:
 4132845
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Shelf Life
  Special Grade
  Application(s)
  Media Suitability
Show More