• USA Home
  • Search Results

  • kromasilcellucoatchiralhplccolumn1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Kromasil® CelluCoat® Chiral HPLC Column

17 Products
 NACRES:
 NB.21
 eCl@ss
 32110501
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Operating parameter
  Matrix active group
  Restriction
  Chromatography Mode
  Application(s)
  Particle Dimension
  Manufacturer designation
  Composite Dimension
Show More

Product #
Description
K08670370 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 4.6 mm,
K08670372 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 4.6 mm,
K08670373 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 4.6 mm,
K08670374 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 4.6 mm,
K08670614 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 10 mm,
K08670615 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 21.2 mm,
K08670616 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 250 mm × 30 mm,
K08670617 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 4.6 mm,
K08670612 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 150 mm × 4.6 mm,
K08670613 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 4.6 mm,
K08670609 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 10 mm,
K08670610 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 21.2 mm,
K08670611 Kromasil® CelluCoat® Chiral HPLC Column, 10 μm particle size, L × I.D. 250 mm × 30 mm,
K08971227 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 50 mm × 2.1 mm,
K08971223 Kromasil® CelluCoat® Chiral HPLC Column, 3 μm particle size, L × I.D. 150 mm × 2.1 mm,
K08971228 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 50 mm × 2.1 mm,
K08971224 Kromasil® CelluCoat® Chiral HPLC Column, 5 μm particle size, L × I.D. 150 mm × 2.1 mm,