• USA Home
  • Search Results

  • l2aminoadipicacid16116111890711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-2-Aminoadipic acid

2 Products
Synonym:   (S)-2-Aminohexanedioic acid, L-Homoglutamic acid, Aad
Image of <SC>L</SC>-2-Aminoadipic acid
CAS Number:
Linear Formula:
 HO2C(CH2)3CH(NH2)CO2H
Molecular Weight:
 161.16
 Beilstein/REAXYS Number:
 1724348
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More