• USA Home
  • Search Results

  • lD2amino4phosphonobutyricacid183102305281511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-(+)-2-Amino-4-phosphonobutyric acid

1 Products
Synonym:   L-AP-4, L-AP4
Image of <SC>L</SC>-<WBR>(+)<WBR>-<WBR>2-<WBR>Amino-<WBR>4-<WBR>phosphonobutyric acid
CAS Number:
Empirical Formula (Hill Notation):
 C4H10NO5P
Molecular Weight:
 183.10
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More