• USA Home
  • Search Results

  • lDlacticacid90087933411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-(+)-Lactic acid

4 Products
Synonym:   (S)-2-Hydroxypropionic acid, Sarcolactic acid
Image of <SC>L</SC>-(+)-Lactic acid
CAS Number:
Empirical Formula (Hill Notation):
 C3H6O3
Molecular Weight:
 90.08
 Beilstein Registry Number:
 1720251
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
  Solubility
  Mode of Action
  Activity Spectrum
Show More