• USA Home
  • Search Results

  • labetalolhydrochloride364873278064611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Labetalol hydrochloride

4 Products
Synonym:   2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzamide hydrochloride
Image of Labetalol hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H24N2O3 · HCl
Molecular Weight:
 364.87
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Solubility
  Manufacturer designation
  Physical Form
Show More