• USA Home
  • Search Results

  • lactoseanhydrous342306342311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lactose (anhydrous)

2 Products
Synonym:   Lactose
Image of Lactose (anhydrous)
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11
Molecular Weight:
 342.30
 Beilstein/REAXYS Number:
 93796
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More