• USA Home
  • Search Results

  • lactosemonohydrate360316404451511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lactose (Monohydrate)

3 Products
Synonym:   β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, D-Lactose monohydrate, Milk sugar
Image of Lactose (Monohydrate)
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11 · H2O
Molecular Weight:
 360.31
 Beilstein/REAXYS Number:
 3768231
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More