• USA Home
  • Search Results

  • lactulose34230461818211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lactulose

7 Products
Synonym:   4-O-β-D-Galactopyranosyl-D-fructofuranose, 4-O-β-D-Galactopyranosyl-D-fructose
Image of Lactulose
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11
Molecular Weight:
 342.30
 Beilstein/REAXYS Number:
 93773
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Anal. Standard Form
  Assay
  Physical Form
  Manufacturer designation
Show More