• USA Home
  • Search Results

  • lalanine89095641711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Alanine

13 Products
Synonym:   (S)-2-Aminopropionic acid, L-α-Aminopropionic acid
Image of <SC>L</SC>-Alanine
CAS Number:
Empirical Formula (Hill Notation):
 C3H7NO2
Molecular Weight:
 89.09
 Beilstein/REAXYS Number:
 1720248
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Allergen
  Organoleptic property
  UV-Absorption Values
  Purity Grade
  Density
  Bulk Density
  pH Value
  Product Line
  Manufacturer designation
  Loss
  Biological Source
  Agency/Method
  Melting Point
  Color
  Physical Form
  Special Grade
  Assay
  Solubility
Show More

Product #
Description
A7627 L-Alanine, ≥98% (TLC),
A7469 L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%,
05129 L-Alanine, BioUltra, ≥99.5% (NT),
A4349 L-Alanine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture,
44526 L-Alanine, certified reference material, TraceCERT®,
05130 L-Alanine, ≥99.0% (NT),
W381829 L-Alanine, ≥99%,
PHR1110 L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material,
1012509 L-Alanine, United States Pharmacopeia (USP) Reference Standard,
W381812 L-Alanine, natural, FG,
PHL80344 L-Alanine, phyproof® Reference Substance,
1.01700 L-Alanine, EMPROVE® EXPERT Ph Eur,USP,
1.01007 L-Alanine, for biochemistry,