• USA Home
  • Search Results

  • lallylglycine115131633848011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Allylglycine

1 Products
Synonym:   (S)-(−)-2-Amino-4-pentenoic acid, L-2-Amino-4-pentenoic acid
Image of <SC>L</SC>-Allylglycine
CAS Number:
Empirical Formula (Hill Notation):
 C5H9NO2
Molecular Weight:
 115.13
 Beilstein/REAXYS Number:
 1721512
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More