• USA Home
  • Search Results

  • lamininfromengelbrethholmswarmmurinesarcomabasementmembrane1234511495681911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane

1 Products
Synonym:   Laminin
CAS Number:
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More