• USA Home
  • Search Results

  • lamotrigine256098405784111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lamotrigine

5 Products
Synonym:   6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X
Image of Lamotrigine
CAS Number:
Empirical Formula (Hill Notation):
 C9H7Cl2N5
Molecular Weight:
 256.09
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Solubility
  Anal. Standard Form
  Physical Form
Show More