• USA Home
  • Search Results

  • lanolin12345800654011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lanolin

3 Products
Synonym:   Wool Grease, Wool Wax, Wool fat
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.25
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
Show More