• USA Home
  • Search Results

  • lanthanumiiibromide378621353679311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lanthanum(III) bromide

1 Products
Synonym:   Lanthanum bromide, Lanthanum tribromide
Image of Lanthanum(III) bromide
CAS Number:
Linear Formula:
 LaBr3
Molecular Weight:
 378.62
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More