• USA Home
  • Search Results

  • lanthanumiiifluoride195901370938111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lanthanum(III) fluoride

1 Products
Image of Lanthanum(III) fluoride
CAS Number:
Linear Formula:
 LaF3
Molecular Weight:
 195.90
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More