• USA Home
  • Search Results

  • larginine13c6hydrochloride2166220174091211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Arginine-13C6 hydrochloride

1 Products
Synonym:   (S)-(+)-2-Amino-5-[(aminomethyl)amino]pentanoic acid-13C6 hydrochloride, 13C Labeled arginine hydrochloride, 13C Labeled arginine, SILAC Amino Acid
Image of <SC>L</SC>-Arginine-<SUP>13</SUP>C<SUB>6</SUB> hydrochloride
CAS Number:
Linear Formula:
 H2N13C(NH)NH13CH2(13CH2)213CH(NH2)13CO2H · HCl
Molecular Weight:
 216.62
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More